5-(Chloromethyl)-3-m-tolyl-[1,2,4]oxadiazole 95+%
Dodavatel: Apollo Scientific
OR346392-1G
OR346392-5G
APOSOR346392-1GEA
6190
CZK
APOSOR346392-1G
APOSOR346392-5G
5-(Chloromethyl)-3-m-tolyl-[1,2,4]oxadiazole 95+%
5-(Chloromethyl)-3-m-tolyl-[1,2,4]oxadiazole
Vzorec:
C₁₀H₉ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD09034175 CAS číslo: 50737-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...