Tisk…

5-Chlorosulphonyl-furan-3-carboxylic acid ethyl ester 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR346347-1G OR346347-5G
APOSOR346347-1GEA 10520 CZK
APOSOR346347-1G APOSOR346347-5G
5-Chlorosulphonyl-furan-3-carboxylic acid ethyl ester 95%
5-Chlorosulphonyl-furan-3-carboxylic acid ethyl ester
Vzorec: C₇H₇ClO₅S
MW: 238,645 g/mol
Bod varu: 118 °C (1 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11109333
CAS číslo: 256373-91-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR