Tisk…

4-Methoxy-benzaldehyde oxime

Dodavatel: Apollo Scientific

OR346329-1G OR346329-5G
APOSOR346329-1GEA 2070 CZK
APOSOR346329-1G APOSOR346329-5G
4-Methoxy-benzaldehyde oxime
4-Methoxy-benzaldehyde oxime
Vzorec: C₈H₉NO₂
MW: 151,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00137433
CAS číslo: 3235-04-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR