Tisk…

4-Hydroxy-5-methoxy-2-nitro-benzoic acid ethyl ester

Dodavatel: Apollo Scientific

OR346322-1G OR346322-5G
APOSOR346322-1GEA 0 CZK
APOSOR346322-1G APOSOR346322-5G
4-Hydroxy-5-methoxy-2-nitro-benzoic acid ethyl ester
4-Hydroxy-5-methoxy-2-nitro-benzoic acid ethyl ester
Vzorec: C₁₀H₁₁NO₆
MW: 241,197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11046180
CAS číslo: 422308-68-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR