Tisk…

3-(3-Methyl-isothiazol-5-yl)-benzaldehyde

Dodavatel: Apollo Scientific

OR346317-500MG
APOSOR346317-500MGEA 9690 CZK
APOSOR346317-500MG
3-(3-Methyl-isothiazol-5-yl)-benzaldehyde
3-(3-Methyl-isothiazol-5-yl)-benzaldehyde
Vzorec: C₁₁H₉NOS
MW: 203,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22573495
CAS číslo: 1401522-01-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR