Tisk…

4-(3-Methyl-isothiazol-5-yl)-benzoic acid

Dodavatel: Apollo Scientific

OR346316-1G OR346316-500MG
APOSOR346316-1GEA 0 CZK
APOSOR346316-1G APOSOR346316-500MG
4-(3-Methyl-isothiazol-5-yl)-benzoic acid
4-(3-Methyl-isothiazol-5-yl)-benzoic acid
Vzorec: C₁₁H₉NO₂S
MW: 219,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18374046
CAS číslo: 1261236-29-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR