4-Bromo-3-methyl-isothiazole-5-carboxylic acid
Dodavatel: Apollo Scientific
OR346312-500MG
OR346312-1G
APOSOR346312-500MGEA
0
CZK
APOSOR346312-500MG
APOSOR346312-1G
4-Bromo-3-methyl-isothiazole-5-carboxylic acid
4-Bromo-3-methyl-isothiazole-5-carboxylic acid
Vzorec:
C₅H₄BrNO₂S MW: 222.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19302667 CAS číslo: 89284-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...