Tisk…
5-Methyl-[1,2,3]thiadiazole-4-carboxylic acid ethyl ester
Dodavatel: Apollo Scientific
OR346278-1G OR346278-500MG
APOSOR346278-1GEA 10910 CZK
APOSOR346278-1G APOSOR346278-500MG
5-Methyl-[1,2,3]thiadiazole-4-carboxylic acid ethyl ester
5-Methyl-[1,2,3]thiadiazole-4-carboxylic acid ethyl ester
Vzorec: C₆H₈N₂O₂S
MW: 172,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100203
CAS číslo: 29682-53-9