Tisk…

Ethylamino-acetic acid ethyl ester

Dodavatel: Apollo Scientific

OR346255-1G OR346255-500MG OR346255-5G
APOSOR346255-1GEA 0 CZK
APOSOR346255-1G APOSOR346255-500MG APOSOR346255-5G
Ethylamino-acetic acid ethyl ester
Ethylamino-acetic acid ethyl ester
Vzorec: C₆H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09953137
CAS číslo: 3183-20-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR