Tisk…

5-(3-Chloro-phenyl)-3-methyl-isoxazole-4-carboxylic acid methyl ester

Dodavatel: Apollo Scientific

OR346224-1G OR346224-5G
APOSOR346224-1GEA 0 CZK
APOSOR346224-1G APOSOR346224-5G
5-(3-Chloro-phenyl)-3-methyl-isoxazole-4-carboxylic acid methyl ester
5-(3-Chloro-phenyl)-3-methyl-isoxazole-4-carboxylic acid methyl ester
Vzorec: C₁₂H₁₀ClNO₃
MW: 251,666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15142852
CAS číslo: 1208081-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR