3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid methyl ester
Dodavatel: Apollo Scientific
OR346223-1G
OR346223-5G
APOSOR346223-1GEA
0
CZK
APOSOR346223-1G
APOSOR346223-5G
3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid methyl ester
3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid methyl ester
Vzorec:
C₁₃H₁₃NO₃ MW: 231,247 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15142851 CAS číslo: 1208081-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...