Tisk…

tert-Butylsulphamoyl chloride

Dodavatel: Apollo Scientific

OR346220-1G OR346220-500MG
APOSOR346220-1GEA 0 CZK
APOSOR346220-1G APOSOR346220-500MG
tert-Butylsulphamoyl chloride
tert-Butylsulphamoyl chloride
Vzorec: C₄H₁₀ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD11520377
CAS číslo: 33581-95-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR