Tisk…

n-Propylsulphamoyl chloride, tech

Dodavatel: Apollo Scientific
Nebezpečí

OR346219-1G OR346219-500MG
APOSOR346219-1GEA 10520 CZK
APOSOR346219-1G APOSOR346219-500MG
n-Propylsulphamoyl chloride, tech
n-Propylsulphamoyl chloride
Vzorec: C₃H₈ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD11520375
CAS číslo: 10305-42-7
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR