Tisk…

2-Chloro-6-methyl-pyridine-3-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR346215-1G OR346215-500MG
APOSOR346215-1GEA 0 CZK
APOSOR346215-1G APOSOR346215-500MG
2-Chloro-6-methyl-pyridine-3-sulphonyl chloride
2-Chloro-6-methyl-pyridine-3-sulphonyl chloride
Vzorec: C₆H₅Cl₂NO₂S
MW: 226,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15142819
CAS číslo: 1208081-60-0
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR