2,5-Dichloro-pyridine-3-sulphonyl chloride
Dodavatel: Apollo Scientific
Synonyms:
2,5-Dichloropyridine-3-sulphonyl chloride
OR346212-1G
OR346212-500MG
OR346212-250MG
APOSOR346212-1GEA
11540
CZK
APOSOR346212-1G
APOSOR346212-500MG
APOSOR346212-250MG
2,5-Dichloro-pyridine-3-sulphonyl chloride
2,5-Dichloro-pyridine-3-sulphonyl chloride
Vzorec:
C₅H₂Cl₃NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD15142815 CAS číslo: 1208081-36-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...