2,3-Dihydro-benzofuran-2-carbonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR346204-1G
OR346204-500MG
APOSOR346204-1GEA
0
CZK
APOSOR346204-1G
APOSOR346204-500MG
2,3-Dihydro-benzofuran-2-carbonyl chloride
2,3-Dihydro-benzofuran-2-carbonyl chloride
Vzorec:
C₉H₇ClO₂ MW: 182,604 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03086169 CAS číslo: 27347-32-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...