2-Chloro-4-methyl-pyridine-3-sulphonic acid amide
Dodavatel: Apollo Scientific
|
Varování
|
OR346200-1G
OR346200-500MG
APOSOR346200-1GEA
0
CZK
APOSOR346200-1G
APOSOR346200-500MG
2-Chloro-4-methyl-pyridine-3-sulphonic acid amide
2-Chloro-4-methyl-pyridine-3-sulphonic acid amide
Vzorec:
C₆H₇ClN₂O₂S MW: 206,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15142805 CAS číslo: 1208081-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...