Tisk…

1-(3-Chloro-phenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR346163-1G OR346163-500MG
APOSOR346163-1GEA 0 CZK
APOSOR346163-1G APOSOR346163-500MG
1-(3-Chloro-phenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
1-(3-Chloro-phenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Vzorec: C₁₂H₁₁ClN₂O₂
MW: 250,681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09934571
CAS číslo: 113808-90-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR