Tisk…

1-m-Tolyl-1H-pyrazole

Dodavatel: Apollo Scientific

OR346160-1G OR346160-5G OR346160-10G
APOSOR346160-1GEA 3540 CZK
APOSOR346160-1G APOSOR346160-5G APOSOR346160-10G
1-m-Tolyl-1H-pyrazole
1-m-Tolyl-1H-pyrazole
Vzorec: C₁₀H₁₀N₂
MW: 158,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11707178
CAS číslo: 850380-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR