2-Chloro-3-chloromethyl-pyridine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-3-(chloromethyl)pyridine
OR346119-5G
APOSOR346119-5GEA
2980
CZK
APOSOR346119-5G
2-Chloro-3-chloromethyl-pyridine 98%
2-Chloro-3-chloromethyl-pyridine
Vzorec:
C₆H₅Cl₂N MW: 162,017 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835107 CAS číslo: 89581-84-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...