Tisk…

5-(tert-Butyloxycarbonyamino)methyl-3-phenyl-[1,2,4]oxadiazole

Dodavatel: Apollo Scientific

OR346074-1G OR346074-5G
APOSOR346074-1GEA 0 CZK
APOSOR346074-1G APOSOR346074-5G
5-(tert-Butyloxycarbonyamino)methyl-3-phenyl-[1,2,4]oxadiazole
5-(tert-Butyloxycarbonyamino)methyl-3-phenyl-[1,2,4]oxadiazole
Vzorec: C₁₄H₁₇N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD10568310
CAS číslo: 1053656-50-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR