Tisk…

1-tert-Butyloxycarbonyl-2-(N-Hydroxycarbamimidoyl)piperidine

Dodavatel: Apollo Scientific

OR346056-1G OR346056-500MG
APOSOR346056-1GEA 0 CZK
APOSOR346056-1G APOSOR346056-500MG
1-tert-Butyloxycarbonyl-2-(N-Hydroxycarbamimidoyl)piperidine
1-tert-Butyloxycarbonyl-2-(N-Hydroxycarbamimidoyl)piperidine
Vzorec: C₁₁H₂₁N₃O₃
MW: 243,303 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568291
CAS číslo: 479080-30-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR