Tisk…

Ethyl 3-(2-tert-butyloxycarbonylaminoethyl)-[1,2,4]oxadiazole-5-carboxylate

Dodavatel: Apollo Scientific

OR346027-1G OR346027-500MG
APOSOR346027-1GEA 0 CZK
APOSOR346027-1G APOSOR346027-500MG
Ethyl 3-(2-tert-butyloxycarbonylaminoethyl)-[1,2,4]oxadiazole-5-carboxylate
Ethyl 3-(2-tert-butyloxycarbonylaminoethyl)-[1,2,4]oxadiazole-5-carboxylate
Vzorec: C₁₂H₁₉N₃O₅
Storage Temperature: Ambient
MDL Number: MFCD10568217
CAS číslo: 1053656-31-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR