Tisk…

Ethyl 5-(4-bromophenyl)-[1,2,4]oxadiazole-3-carboxylate

Dodavatel: Apollo Scientific
Varování

OR346026-1G OR346026-5G
APOSOR346026-1GEA 0 CZK
APOSOR346026-1G APOSOR346026-5G
Ethyl 5-(4-bromophenyl)-[1,2,4]oxadiazole-3-carboxylate
Ethyl 5-(4-bromophenyl)-[1,2,4]oxadiazole-3-carboxylate
Vzorec: C₁₁H₉BrN₂O₃
MW: 297,105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568214
CAS číslo: 1053656-27-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR