Tisk…

5-Aminopyrimidin-4(3H)-one

Dodavatel: Apollo Scientific

OR345717-1G OR345717-250MG
APOSOR345717-1GEA 6250 CZK
APOSOR345717-1G APOSOR345717-250MG
5-Aminopyrimidin-4(3H)-one
5-Aminopyrimidin-4(3H)-one
Vzorec: C₄H₅N₃O
MW: 111,102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11574230
CAS číslo: 69785-94-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR