Tisk…

4-Methoxypyridazine 97%

Dodavatel: Apollo Scientific

OR345702-1G
APOSOR345702-1GEA 3020 CZK
APOSOR345702-1G
4-Methoxypyridazine 97%
4-Methoxypyridazine
Vzorec: C₅H₆N₂O
MW: 110,114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236840
CAS číslo: 20733-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR