4-Amino-2-chloro-3-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloro-3-methylpyridin-4-amine
OR345541-1G
OR345541-250MG
OR345541-5G
APOSOR345541-1GEA
4530
CZK
APOSOR345541-1G
APOSOR345541-250MG
APOSOR345541-5G
4-Amino-2-chloro-3-methylpyridine
4-Amino-2-chloro-3-methylpyridine
Vzorec:
C₆H₇ClN₂ MW: 142,586 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18415428 CAS číslo: 79055-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...