Tisk…

2-Hydrazino-5-iodopyridine

Dodavatel: Apollo Scientific
Varování

OR345503-1G OR345503-250MG
APOSOR345503-1GEA 16340 CZK
APOSOR345503-1G APOSOR345503-250MG
2-Hydrazino-5-iodopyridine
2-Hydrazino-5-iodopyridine
Vzorec: C₅H₆IN₃
MW: 235,026 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09999999
CAS číslo: 77992-46-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR