2-Bromo-6-methoxy-3-nitropyridine
Dodavatel: Apollo Scientific
OR345495-5G
APOSOR345495-5GEA
1660
CZK
APOSOR345495-5G
2-Bromo-6-methoxy-3-nitropyridine
2-Bromo-6-methoxy-3-nitropyridine
Vzorec:
C₆H₅BrN₂O₃ MW: 233,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07774106 CAS číslo: 344296-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...