Tisk…

2-Bromo-3-chloro-4-methylpyridine 98%

Dodavatel: Apollo Scientific

OR345441-5G
APOSOR345441-5GEA 2030 CZK
APOSOR345441-5G
2-Bromo-3-chloro-4-methylpyridine 98%
2-Bromo-3-chloro-4-methylpyridine
Vzorec: C₆H₅BrClN
MW: 206,468 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08277283
CAS číslo: 884495-42-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR