Tisk…

5-Chloro-2-methylpyridine 97%

Dodavatel: Apollo Scientific

OR345400-25G
APOSOR345400-25GEA 4140 CZK
APOSOR345400-25G
5-Chloro-2-methylpyridine 97%
5-Chloro-2-methylpyridine
Vzorec: C₆H₆ClN
MW: 127,572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114188
CAS číslo: 72093-07-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR