Tisk…

5-Methoxy-2-methylpyridine 95%

Dodavatel: Apollo Scientific
Varování

OR345367-1G OR345367-5G
APOSOR345367-1GEA 3770 CZK
APOSOR345367-1G APOSOR345367-5G
5-Methoxy-2-methylpyridine 95%
5-Methoxy-2-methylpyridine
Vzorec: C₇H₉NO
MW: 123.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09833595
CAS číslo: 55270-47-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR