5-Amino-2-cyano-3-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-3-methylpicolinonitrile
OR345307-1G
OR345307-5G
APOSOR345307-1GEA
4070
CZK
APOSOR345307-1G
APOSOR345307-5G
5-Amino-2-cyano-3-methylpyridine
5-Amino-2-cyano-3-methylpyridine
Vzorec:
C₇H₇N₃ MW: 133.151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375122 CAS číslo: 252056-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...