2-Bromo-6-chloro-4-methylpyridine 98%
Dodavatel: Apollo Scientific
OR345269-1G
OR345269-10G
OR345269-5G
APOSOR345269-1GEA
2440
CZK
APOSOR345269-1G
APOSOR345269-10G
APOSOR345269-5G
2-Bromo-6-chloro-4-methylpyridine 98%
2-Bromo-6-chloro-4-methylpyridine
Vzorec:
C₆H₅BrClN MW: 206,468 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09839268 CAS číslo: 157329-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...