Tisk…

3,4-Dichloro-2-methylpyridine 95%

Dodavatel: Apollo Scientific

OR345182-1G OR345182-250MG
APOSOR345182-1GEA 8700 CZK
APOSOR345182-1G APOSOR345182-250MG
3,4-Dichloro-2-methylpyridine 95%
3,4-Dichloro-2-methylpyridine
Vzorec: C₆H₅Cl₂N
MW: 162,017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839289
CAS číslo: 103949-58-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR