Tisk…

2-Chloro-1-naphthol 97%

Dodavatel: Apollo Scientific
Varování

OR34495-1G OR34495-250MG OR34495-5G
APOSOR34495-1GEA 13560 CZK
APOSOR34495-1G APOSOR34495-250MG APOSOR34495-5G
2-Chloro-1-naphthol 97%
2-Chloro-1-naphthol
Vzorec: C₁₀H₇ClO
MW: 178,615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415868
CAS číslo: 606-40-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR