Tisk…

4-Bromo-N,N-dimethylbenzylamine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(4-Bromophenyl)-N,N-dimethylmethanamine, 1-(4-Bromophenyl)-N,N-dimethylmethylamine

OR3370-10G OR3370-1G OR3370-5G
APOSOR3370-10GEA 7670 CZK
APOSOR3370-10G APOSOR3370-1G APOSOR3370-5G
4-Bromo-N,N-dimethylbenzylamine 98%
4-Bromo-N,N-dimethylbenzylamine
Vzorec: C₉H₁₂BrN
MW: 214,102 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00032473
CAS číslo: 6274-57-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR