Tisk…

2-Hydrazino-4-iodopyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydrazinyl-4-iodopyridine

OR33683-1G OR33683-250MG OR33683-5G
APOSOR33683-1GEA 3740 CZK
APOSOR33683-1G APOSOR33683-250MG APOSOR33683-5G
2-Hydrazino-4-iodopyridine
2-Hydrazino-4-iodopyridine
Vzorec: C₅H₆IN₃
MW: 235,026 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10000014
CAS číslo: 1057393-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR