Tisk…

3-Bromo-5-chloro-2-hydrazinopyridine

Dodavatel: Apollo Scientific
Varování

OR33678-1G OR33678-250MG
APOSOR33678-1GEA 1240 CZK
APOSOR33678-1G APOSOR33678-250MG
3-Bromo-5-chloro-2-hydrazinopyridine
3-Bromo-5-chloro-2-hydrazinopyridine
Vzorec: C₅H₅BrClN₃
MW: 222,47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10000080
CAS číslo: 1289113-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR