Tisk…

2-Hydrazino-3-iodopyridine

Dodavatel: Apollo Scientific
Varování

OR33677-1G OR33677-250MG OR33677-5G
APOSOR33677-1GEA 13230 CZK
APOSOR33677-1G APOSOR33677-250MG APOSOR33677-5G
2-Hydrazino-3-iodopyridine
2-Hydrazino-3-iodopyridine
Vzorec: C₅H₆IN₃
MW: 235,026 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10000048
CAS číslo: 54231-42-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR