Tisk…

2-Amino-6-methoxyphenol 95%

Dodavatel: Apollo Scientific
Varování

OR33674-1G OR33674-250MG
APOSOR33674-1GEA 17360 CZK
APOSOR33674-1G APOSOR33674-250MG
2-Amino-6-methoxyphenol 95%
2-Amino-6-methoxyphenol
Vzorec: C₇H₉NO₂
MW: 139.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16998784
CAS číslo: 40925-71-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR