Tisk…

2-Methyl-1-(4-methylpiperazin-1-yl)propan-2-amine 95%

Dodavatel: Apollo Scientific
Varování

OR33669-1G OR33669-5G
APOSOR33669-1GEA 13460 CZK
APOSOR33669-1G APOSOR33669-5G
2-Methyl-1-(4-methylpiperazin-1-yl)propan-2-amine 95%
2-Methyl-1-(4-methylpiperazin-1-yl)propan-2-amine
Vzorec: C₉H₂₁N₃
MW: 171,283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06751854
CAS číslo: 84725-48-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR