Tisk…

5-Chlorobenzofuran-2-carboxaldehyde

Dodavatel: Apollo Scientific
Varování

OR33663-1G OR33663-250MG
APOSOR33663-1GEA 7410 CZK
APOSOR33663-1G APOSOR33663-250MG
5-Chlorobenzofuran-2-carboxaldehyde
5-Chlorobenzofuran-2-carboxaldehyde
Vzorec: C₉H₅ClO₂
MW: 180,588 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11574566
CAS číslo: 23145-14-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR