Tisk…

(1-Cyclopropylpyrazol-3-yl)methanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR33662-100MG OR33662-1G OR33662-250MG
APOSOR33662-100MGEA 910 CZK
APOSOR33662-100MG APOSOR33662-1G APOSOR33662-250MG
(1-Cyclopropylpyrazol-3-yl)methanamine hydrochloride
(1-Cyclopropylpyrazol-3-yl)methanamine hydrochloride
Vzorec: C₇H₁₁N₃·ClH
MW: 173,643 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31812083
CAS číslo: 2391987-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR