Tisk…

(2E)-3-(3-Iodophenyl)prop-2-enoic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 3-(3-Iodophenyl)acrylic acid

OR33644-1G OR33644-5G
APOSOR33644-1GEA 3150 CZK
APOSOR33644-1G APOSOR33644-5G
(2E)-3-(3-Iodophenyl)prop-2-enoic acid 97%
(2E)-3-(3-Iodophenyl)prop-2-enoic acid
Vzorec: C₉H₇IO₂
Storage Temperature: Ambient
MDL Number: MFCD00464508
CAS číslo: 41070-12-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR