Tisk…

5-Bromo-3-furoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromofuran-3-carboxylic acid, 2-Bromo-4-carboxyfuran

OR3279-1G OR3279-500MG OR3279-5G
APOSOR3279-1GEA 1370 CZK
APOSOR3279-1G APOSOR3279-500MG APOSOR3279-5G
5-Bromo-3-furoic acid
5-Bromo-3-furoic acid
Vzorec: C₅H₃BrO₃
MW: 190,98 g/mol
Bod tání: 130…139 °C
Storage Temperature: Ambient
MDL Number: MFCD08669547
CAS číslo: 58832-36-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR