Tisk…

5-Iodo-2-(1H-pyrrol-1-yl)pyridine 95%

Dodavatel: Apollo Scientific

OR32766-1G OR32766-5G
APOSOR32766-1GEA 5790 CZK
APOSOR32766-1G APOSOR32766-5G
5-Iodo-2-(1H-pyrrol-1-yl)pyridine 95%
5-Iodo-2-(1H-pyrrol-1-yl)pyridine
Vzorec: C₉H₇IN₂
MW: 270,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01315196
CAS číslo: 338748-93-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR