Methyl 4-(morpholin-4-yl)benzoate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 4-morpholinobenzoate
OR32672-1G
APOSOR32672-1GEA
5130
CZK
APOSOR32672-1G
Methyl 4-(morpholin-4-yl)benzoate 95%
Methyl 4-(morpholin-4-yl)benzoate
Vzorec:
C₁₂H₁₅NO₃ MW: 221,252 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03012127 CAS číslo: 23676-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...