Tisk…

(2Z)-2-(4-Chlorophenyl)-3-(dimethylamino)prop-2-enal 95%

Dodavatel: Apollo Scientific

OR32628-1G OR32628-5G
APOSOR32628-1GEA 7670 CZK
APOSOR32628-1G APOSOR32628-5G
(2Z)-2-(4-Chlorophenyl)-3-(dimethylamino)prop-2-enal 95%
(2Z)-2-(4-Chlorophenyl)-3-(dimethylamino)prop-2-enal
Vzorec: C₁₁H₁₂ClNO
MW: 209,672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973720
CAS číslo: 19927-64-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR