6-Methyl-2-(pyridin-3-yl)pyrimidin-4-ol 95%
Dodavatel: Apollo Scientific
OR32546-1G
OR32546-5G
APOSOR32546-1GEA
9660
CZK
APOSOR32546-1G
APOSOR32546-5G
6-Methyl-2-(pyridin-3-yl)pyrimidin-4-ol 95%
6-Methyl-2-(pyridin-3-yl)pyrimidin-4-ol
Vzorec:
C₁₀H₉N₃O MW: 187,198 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00113965 CAS číslo: 16879-57-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...